Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014391.2 | NP_055206.2 | 2 | 334 | ||
GenBank mRNA | AK290960.1 | - | 2 | 146 | ||
AU139209.1 | - | 2 | 336 | |||
AY903446.1 | - | 2 | 103 | |||
BC005191.1 | - | 2 | 147 | |||
BC018667.1 | - | 2 | 150 | 4245591 | ||
X83703.1 | - | 2 | 335 |