Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164380.1 | NP_001157852.1 | 5 | 512 | ||
NM_001164381.1 | NP_001157853.1 | 4 | 381 | |||
NM_001164384.1 | NP_001157856.1 | 3 | 445 | |||
NM_001164385.1 | NP_001157857.1 | 2 | 202 | |||
NM_014393.2 | NP_055208.2 | 4 | 381 | |||
GenBank mRNA | AB210014.1 | - | 4 | 277 | ||
AF459097.1 | - | 3 | 231 | |||
AF459098.1 | - | 2 | 100 | |||
AK002152.1 | - | 5 | 425 | |||
AK290141.1 | - | 3 | 445 | |||
AK293729.1 | - | 2 | 142 | |||
AK294466.1 | - | 2 | 154 | |||
AK314617.1 | - | 4 | 294 | |||
AL079285.1 | - | 4 | 304 | |||
BC008369.1 | - | 4 | 294 | 3682739 | ||
BC008370.1 | - | 4 | 294 | |||
BC110447.1 | - | 2 | 118 | 40009098 | ||
DB501161.1 | - | 2 | 202 | |||
Y19062.1 | - | 4 | 276 |