Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014400.2 | NP_055215.2 | 5 | 1371 | ||
GenBank mRNA | AF082889.1 | - | 5 | 1348 | ||
AJ223603.2 | - | 5 | 1299 | |||
AK075464.1 | - | 5 | 1300 | |||
AY359006.1 | - | 5 | 1278 | |||
BC039167.1 | - | 5 | 1285 | 4751561 | ||
BC041061.1 | - | 5 | 1280 |