Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175039.3 | NP_778204.1 | 6 | 1460 | ||
NM_175040.3 | NP_778205.1 | 5 | 1373 | |||
XM_011518527.2 | XP_011516829.1 | 7 | 1552 | |||
XM_011518529.2 | XP_011516831.1 | 7 | 1583 | |||
GenBank mRNA | AK000600.1 | - | 7 | 1411 | ||
AK292049.1 | - | 6 | 1215 | |||
BC036705.2 | - | 6 | 1363 | 4309208 | ||
F27902.1 | - | 1 | 236 |