Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032172.2 | NP_115548.1 | 2 | 325 | ||
XM_005249883.4 | XP_005249940.1 | 2 | 272 | |||
XM_006715790.1 | XP_006715853.1 | 2 | 272 | |||
XM_006715791.2 | XP_006715854.1 | 5 | 778 | |||
XM_011515573.1 | XP_011513875.1 | 6 | 837 | |||
XM_011515574.1 | XP_011513876.1 | 2 | 270 | |||
XM_011515578.1 | XP_011513880.1 | 2 | 267 | |||
GenBank mRNA | AJ601395.1 | - | 1 | 202 | ||
AK022759.1 | - | 2 | 269 | |||
AK298164.1 | - | 2 | 296 | |||
AK309375.1 | - | 2 | 267 | |||
AY618868.1 | - | 2 | 269 | |||
BC060846.1 | - | 2 | 264 | |||
BC132862.1 | - | 1 | 222 | 40146884 | ||
BC136656.1 | - | 1 | 222 | |||
DR001777.1 | - | 2 | 329 |