Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000171.3 | NP_000162.2 | 7, 8 | 1207 | ||
NM_001146040.1 | NP_001139512.1 | 7, 8 | 1207 | |||
NM_001292000.1 | NP_001278929.1 | 6, 7 | 1079 | |||
XM_017009349.1 | XP_016864838.1 | 7, 8 | 968 | |||
GenBank mRNA | AK226046.1 | - | 6, 7 | 1079 | ||
AK312702.1 | - | 7, 8 | 1133 | |||
BC074980.2 | - | 7, 8 | 961 | 30915298 | ||
BC114947.1 | - | 7, 8 | 1207 | 40084102 | ||
BC114967.1 | - | 6, 7 | 1079 | 40084103 | ||
X52009.1 | - | 7, 8 | 1216 |