Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000171.3 | NP_000162.2 | 9 | 1639 | ||
NM_001146040.1 | NP_001139512.1 | 9 | 1663 | |||
NM_001292000.1 | NP_001278929.1 | 8 | 1511 | |||
XM_017009349.1 | XP_016864838.1 | 9 | 1400 | |||
GenBank mRNA | BC074980.2 | - | 9 | 1393 | 30915298 | |
BC114947.1 | - | 9 | 1663 | 40084102 | ||
BC114967.1 | - | 8 | 1511 | 40084103 | ||
X52009.1 | - | 9 | 1648 |