Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000285.3 | NP_000276.2 | 15 | 1759 | ||
NM_001166056.1 | NP_001159528.1 | 13 | 1636 | |||
NM_001166057.1 | NP_001159529.1 | 13 | 1567 | |||
GenBank mRNA | AK057538.1 | - | 2 | 1493 | ||
AK225037.1 | - | 15 | 1640 | |||
AK290756.1 | - | 15 | 1640 | |||
AK290781.1 | - | 15 | 1655 | |||
AK291561.1 | - | 15 | 1657 | |||
AK294619.1 | - | 13 | 1534 | |||
BC004305.1 | - | 15 | 1630 | 3626134 | ||
BC015027.1 | - | 15 | 1660 | 3920275 | ||
BC028295.1 | - | 15 | 1631 | 4470066 | ||
J04605.1 | - | 15 | 1642 |