Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000774.4 | NP_000765.2 | 9 | 1313 | ||
NR_135528.1 | 9 | 1255 | ||||
XM_011526551.2 | XP_011524853.1 | 9 | 1453 | |||
XM_011526552.2 | XP_011524854.1 | 9 | 1456 | |||
XM_011526554.2 | XP_011524856.1 | 8 | 1271 | |||
XM_017026384.1 | XP_016881873.1 | 8 | 1644 | |||
XM_017026385.1 | XP_016881874.1 | 8 | 1644 | |||
GenBank mRNA | AK298082.1 | - | 8 | 1844 | ||
AK316498.1 | - | 9 | 1255 | |||
BC109056.1 | - | 9 | 1232 | 40033365 | ||
J02906.1 | - | 9 | 1296 |