Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000883.3 | NP_000874.2 | 14 | 1618 | ||
NM_001102605.1 | NP_001096075.1 | 13 | 1588 | |||
NM_001142573.1 | NP_001136045.1 | 11 | 1388 | |||
NM_001142574.1 | NP_001136046.1 | 11 | 1373 | |||
NM_001142575.1 | NP_001136047.1 | 10 | 1313 | |||
NM_001142576.1 | NP_001136048.1 | 13 | 1519 | |||
NM_001304521.1 | NP_001291450.1 | 11 | 1416 | |||
NM_183243.2 | NP_899066.1 | 12 | 1510 | |||
XM_017012168.1 | XP_016867657.1 | 13 | 1588 | |||
XM_017012169.1 | XP_016867658.1 | 12 | 1510 | |||
XM_017012170.1 | XP_016867659.1 | 11 | 1417 | |||
XM_017012171.1 | XP_016867660.1 | 13 | 1706 | |||
XM_017012172.1 | XP_016867661.1 | 13 | 1706 | |||
GenBank mRNA | AK054640.1 | - | 10 | 1285 | ||
AK054667.1 | - | 12 | 1510 | |||
AK092452.1 | - | 13 | 1555 | |||
AK122994.1 | - | 13 | 1587 | |||
AK131294.1 | - | 11 | 1335 | |||
AK293413.1 | - | 11 | 1345 | |||
AK297943.1 | - | 11 | 1360 | |||
BC033622.1 | - | 12 | 1501 | 5210847 | ||
CD014008.1 | - | 12 | 1302 | |||
CD014010.1 | - | 13 | 1424 | |||
CR933672.1 | - | 1554 | ||||
J05272.1 | - | 14 | 1882 |
Gene Symbol: IMPDH1P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC127892.1 | - | 1 | 76 |