Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003463.4 | NP_003454.1 | 6 | 2349 | ||
XM_011536111.1 | XP_011534413.1 | 6 | 1688 | |||
XM_011536112.1 | XP_011534414.1 | 6 | 2447 | |||
XM_017011270.1 | XP_016866759.1 | 6 | 1705 | |||
XM_017011271.1 | XP_016866760.1 | 6 | 1685 | |||
GenBank mRNA | BC023975.2 | - | 6 | 1791 | 2960001 | |
CR749458.1 | - | 8 | 2182 | |||
U48296.1 | - | 5 | 1587 |