Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282236.1 | NP_001269165.1 | 2 | 779 | ||
NM_004196.4 | NP_004187.2 | 1 | 187 | |||
XM_005268157.2 | XP_005268214.1 | 2 | 779 | |||
XM_005268159.2 | XP_005268216.1 | 2 | 779 | |||
XM_017021729.1 | XP_016877218.1 | 2 | 671 | |||
XM_017021730.1 | XP_016877219.1 | 2 | 671 | |||
XM_017021731.1 | XP_016877220.1 | 2 | 680 | |||
XM_017021732.1 | XP_016877221.1 | 2 | 671 | |||
GenBank mRNA | AF390028.1 | - | 5 | 701 | ||
AK308732.1 | - | 2 | 774 | |||
BC104977.1 | - | 1 | 162 | 8143980 | ||
X66358.1 | - | 1 | 347 |