Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184705.2 | NP_001171634.2 | 4 | 746 | ||
NM_005327.4 | NP_005318.3 | 4 | 746 | |||
XM_005262972.2 | XP_005263029.1 | 4 | 678 | |||
GenBank mRNA | AF001902.1 | - | 4 | 510 | ||
AF001903.1 | - | 4 | 746 | |||
AK096018.1 | - | 4 | 678 | |||
AK314444.1 | - | 4 | 607 | |||
BC000306.2 | - | 4 | 554 | 2820507 | ||
X96752.1 | - | 4 | 614 |