Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRG1 SIRNA Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location:
Chr.16: 57619535 - 57665039 on Build GRCh38
UniGene ID:
Hs.513633
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
212773
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145770.2
NP_001139242.1
14
2090
NM_001145771.2
NP_001139243.1
14
2546
NM_001145772.2
NP_001139244.1
14
2554
NM_001145773.2
NP_001139245.1
13
2425
NM_001145774.2
NP_001139246.1
14
2327
NM_001290142.1
NP_001277071.1
13
1648
NM_001290143.1
NP_001277072.1
13
1997
NM_001290144.1
NP_001277073.1
13
2166
NM_005682.6
NP_005673.3
14
2108
NM_201524.3
NP_958932.1
14
2532
NM_201525.3
NP_958933.1
13
2410
XM_005256237.2
XP_005256294.1
13
2144
XM_005256238.2
XP_005256295.1
14
2483
XM_005256239.2
XP_005256296.1
14
2245
XM_005256240.4
XP_005256297.1
13
1966
XM_005256241.4
XP_005256298.1
13
1970
XM_005256242.4
XP_005256299.1
13
2027
XM_005256244.3
XP_005256301.1
13
2119
XM_005256245.2
XP_005256302.1
13
2170
XM_005256246.2
XP_005256303.1
14
2230
XM_005256247.2
XP_005256304.1
13
2129
XM_005256248.2
XP_005256305.1
13
2194
XM_005256249.3
XP_005256306.1
13
1985
XM_005256251.4
XP_005256308.1
13
2011
XM_005256252.2
XP_005256309.1
13
1955
XM_005256254.2
XP_005256311.1
13
2155
XM_005256255.2
XP_005256312.1
14
2450
XM_006721338.2
XP_006721401.1
14
2266
XM_006721339.3
XP_006721402.1
14
2073
XM_006721340.3
XP_006721403.1
14
2331
XM_006721341.2
XP_006721404.1
14
2288
XM_006721342.2
XP_006721405.1
14
2251
XM_006721343.3
XP_006721406.1
14
2316
XM_006721344.2
XP_006721407.1
14
2273
XM_006721345.3
XP_006721408.1
14
2042
XM_006721346.2
XP_006721409.1
13
1897
XM_006721347.2
XP_006721410.1
14
2465
XM_011523461.2
XP_011521763.1
14
2234
XM_011523462.2
XP_011521764.1
13
2049
XM_011523463.2
XP_011521765.1
14
2059
XM_011523464.2
XP_011521766.1
14
2219
XM_011523465.2
XP_011521767.1
14
2157
XM_011523466.2
XP_011521768.1
13
2204
XM_011523467.2
XP_011521769.1
14
1951
XM_011523468.2
XP_011521770.1
13
1953
XM_017023892.1
XP_016879381.1
13
2137
GenBank mRNA
AF106858.1
-
13
1978
AJ011001.1
-
14
2114
AK075270.1
-
14
2042
AK075555.1
-
14
2065
AK131550.1
-
13
1603
AK297236.1
-
13
2039
AK299110.1
-
13
1954
AK311156.1
-
3
384
AK312405.1
-
13
2246
AY358400.1
-
14
2029
BC008770.2
-
13
1974
3139174
BC013207.1
-
14
2104
BT007311.1
-
12
1816
CR936747.1
-
13
1939
EU432119.1
-
12
1816
KU178472.1
-
12
1816
KU178473.1
-
12
1798
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cell migration
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
heparin binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-fls9w:80/100.66.78.88:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline