Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005990.3 | NP_005981.3 | 2, 3 | 623 | ||
XM_017009788.1 | XP_016865277.1 | 1, 2 | 177 | |||
GenBank mRNA | AB015718.1 | - | 2, 3 | 373 | ||
AK307943.1 | - | 2, 3 | 157 | |||
AK313350.1 | - | 3, 4 | 378 | |||
BC070077.1 | - | 2, 3 | 537 | 5267449 | ||
BI459912.1 | - | 2, 3 | 542 |