Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006343.2 | NP_006334.2 | 2 | 381 | ||
XM_005263565.4 | XP_005263622.1 | 2 | 516 | |||
XM_005263568.4 | XP_005263625.1 | 2 | 516 | |||
XM_011510490.2 | XP_011508792.1 | 2 | 416 | |||
XM_017003164.1 | XP_016858653.1 | 2 | 386 | |||
XM_017003165.1 | XP_016858654.1 | 2 | 433 | |||
GenBank mRNA | AK316584.1 | - | 2 | 388 | ||
BX474860.1 | - | 2 | 382 | |||
U08023.1 | - | 2 | 396 |