Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190799.1 | NP_001177728.1 | 4 | 1286 | ||
NM_001190801.1 | NP_001177730.1 | 5 | 1205 | |||
NM_014264.4 | NP_055079.3 | 5 | 1382 | |||
XM_005262701.2 | XP_005262758.1 | 5 | 1344 | |||
XM_017007662.1 | XP_016863151.1 | 5 | 1541 | |||
XM_017007663.1 | XP_016863152.1 | 5 | 1542 | |||
GenBank mRNA | AB006972.1 | - | 5 | 1296 | ||
AK302858.1 | - | 5 | 1205 | |||
AK303399.1 | - | 4 | 1220 | |||
AK314238.1 | - | 5 | 1350 | |||
BC036023.1 | - | 5 | 1350 | 5273226 | ||
Y13115.1 | - | 5 | 1248 |