Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306168.1 | NP_001293097.1 | 2 | 921 | ||
NM_020396.3 | NP_065129.1 | 2 | 851 | |||
GenBank mRNA | AF326964.1 | - | 2 | 851 | ||
BC104442.1 | - | 2 | 845 | 40022820 | ||
BC104443.1 | - | 2 | 845 | 40022822 |