Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145155.1 | NP_001138627.1 | 2 | 744 | ||
NM_001145156.1 | NP_001138628.1 | 2 | 376 | |||
NM_001145157.1 | NP_001138629.1 | 2 | 837 | |||
NM_021005.3 | NP_066285.1 | 2 | 1985 | |||
GenBank mRNA | AB307711.1 | - | 2 | 761 | ||
AK298824.1 | - | 2 | 424 | |||
AK301595.1 | - | 2 | 381 | |||
AK316086.1 | - | 2 | 376 | |||
BC014664.1 | - | 2 | 911 | 4849984 | ||
BC034585.1 | - | 2 | 1014 | |||
BC042897.1 | - | 3 | 1342 | 5177487 | ||
BC106083.1 | - | 2 | 744 | 4453629 | ||
FM208183.1 | - | 2 | 734 | |||
HQ692848.1 | - | 2 | 761 | |||
HQ692849.1 | - | 2 | 362 | |||
HQ692850.1 | - | 1 | 302 | |||
M62760.1 | - | 2 | 1974 | |||
M64497.1 | - | 2 | 1103 |