Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033274.4 | NP_150377.1 | 15 | 1803 | ||
XM_005266003.2 | XP_005266060.1 | 15 | 1755 | |||
XM_011534682.2 | XP_011532984.1 | 8 | 2559 | |||
XM_017010009.1 | XP_016865498.1 | 8 | 1020 | |||
GenBank mRNA | AF311317.1 | - | 15 | 1712 | ||
AK091464.1 | - | 15 | 1765 | |||
AK308182.1 | - | 15 | 1755 | |||
BC024214.2 | - | 4 | 388 | |||
BC033132.1 | - | 4 | 388 | |||
Y13786.2 | - | 15 | 1753 |