Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320586.1 | NP_001307515.1 | 6 | 573 | ||
NM_001320587.1 | NP_001307516.1 | 5 | 480 | |||
NM_001320588.1 | NP_001307517.1 | 3 | 274 | |||
NM_001320589.1 | NP_001307518.1 | 3 | 525 | |||
NM_001320590.1 | NP_001307519.1 | 3 | 525 | |||
NM_138448.3 | NP_612457.1 | 3 | 525 | |||
XM_017005406.1 | XP_016860895.1 | 7 | 919 | |||
XM_017005407.1 | XP_016860896.1 | 6 | 838 | |||
XM_017005408.1 | XP_016860897.1 | 5 | 1547 | |||
XM_017005409.1 | XP_016860898.1 | 5 | 745 | |||
XM_017005410.1 | XP_016860899.1 | 3 | 395 | |||
XM_017005411.1 | XP_016860900.1 | 7 | 919 | |||
XM_017005412.1 | XP_016860901.1 | 7 | 919 | |||
XM_017005413.1 | XP_016860902.1 | 7 | 919 | |||
GenBank mRNA | BC012290.1 | - | 3 | 294 | 3864319 | |
BF671963.1 | - | 3 | 273 | |||
BG741807.1 | - | 5 | 474 | |||
BU543943.1 | - | 6 | 573 | |||
BU857820.1 | - | 3 | 383 | |||
X84195.1 | - | 3 | 274 |