Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206804.1 | NP_001193733.1 | 16 | 1099 | ||
NM_002757.3 | NP_002748.1 | 16 | 1683 | |||
NM_145160.2 | NP_660143.1 | 16 | 1683 | |||
GenBank mRNA | AK024766.1 | - | 5 | 277 | ||
AK057521.1 | - | 10 | 811 | |||
AK293459.1 | - | 16 | 1099 | |||
AK315435.1 | - | 16 | 1230 | |||
BC008838.2 | - | 16 | 1340 | 4100679 | ||
BT006780.1 | - | 16 | 1030 | |||
CR542229.1 | - | 16 | 1030 | |||
U71087.1 | - | 16 | 1091 | |||
U71088.2 | - | 16 | 1035 |