Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014453.3 | NP_055268.1 | 2 | 286 | ||
NM_198426.2 | NP_940818.1 | 2 | 288 | |||
XM_005258746.2 | XP_005258803.1 | 2 | 786 | |||
XM_005258747.3 | XP_005258804.1 | 2 | 909 | |||
GenBank mRNA | AF042384.1 | - | 2 | 254 | ||
AK092868.1 | - | 1 | 925 | |||
AK311974.1 | - | 2 | 174 | |||
AY364248.1 | - | 1 | 125 | |||
BC002502.2 | - | 2 | 258 | 3051024 | ||
BG546479.1 | - | 2 | 282 | |||
BI457978.1 | - | 2 | 278 | |||
BP315111.1 | - | 2 | 286 | |||
BT007298.1 | - | 1 | 125 | |||
CN305066.1 | - | 2 | 289 | |||
CR457002.1 | - | 1 | 125 |