Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184763.1 | NP_001171692.1 | 2 | 295 | ||
NM_144624.2 | NP_653225.2 | 2 | 636 | |||
NM_175866.4 | NP_787062.1 | 2 | 636 | |||
GenBank mRNA | AJ536197.1 | - | 2 | 585 | ||
AK058195.1 | - | 2 | 295 | |||
AK292369.1 | - | 2 | 459 | |||
AL834136.1 | - | 2 | 356 | |||
BC014917.1 | - | 1 | 125 | |||
BC026046.1 | - | 2 | 598 | 4649261 |