Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178170.2 | NP_835464.1 | 15 | 2260 | ||
XM_011524638.2 | XP_011522940.1 | 16 | 2503 | |||
XM_011524640.2 | XP_011522942.1 | 16 | 2464 | |||
XM_017024499.1 | XP_016879988.1 | 15 | 2309 | |||
XM_017024500.1 | XP_016879989.1 | 15 | 2345 | |||
GenBank mRNA | AK301429.1 | - | 11 | 1794 | ||
AL833909.1 | - | 6 | 930 | |||
AY242354.1 | - | 15 | 2260 |