Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000754.3 | NP_000745.1 | 5 | 782 | ||
NM_001135161.1 | NP_001128633.1 | 5 | 740 | |||
NM_001135162.1 | NP_001128634.1 | 5 | 757 | |||
NM_007310.2 | NP_009294.1 | 3 | 513 | |||
XM_011529886.1 | XP_011528188.1 | 5 | 687 | |||
XM_017028594.1 | XP_016884083.1 | 5 | 1006 | |||
XM_017028595.1 | XP_016884084.1 | 5 | 950 | |||
GenBank mRNA | AK130031.1 | - | 6 | 562 | ||
AK290440.1 | - | 5 | 720 | |||
BC000419.2 | - | 5 | 710 | 2964400 | ||
BC005867.2 | - | 5 | 710 | 2964400 | ||
BC011935.2 | - | 4 | 625 | 3503220 | ||
BC100018.1 | - | 5 | 772 | 6163659 | ||
BX461394.2 | - | 5 | 790 | |||
CR456422.1 | - | 4 | 548 | |||
CR456997.1 | - | 3 | 533 | |||
EF103918.1 | - | 3 | 274 | |||
EF103922.1 | - | 4 | 562 | |||
FJ224345.1 | - | 5 | 710 | |||
FJ410131.1 | - | 3 | 383 | |||
FJ410132.1 | - | 3 | 533 | |||
M58525.1 | - | 5 | 737 | |||
M65212.1 | - | 4 | 611 |