Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 8 | 1456 | ||
NM_001099773.1 | NP_001093243.1 | 8 | 1530 | |||
GenBank mRNA | AK056794.1 | - | 8 | 1525 | ||
AK225715.1 | - | 7 | 1044 | |||
AK292300.1 | - | 8 | 1362 | |||
AK292301.1 | - | 7 | 1320 | |||
AK293211.1 | - | 9 | 1322 | |||
AK300178.1 | - | 8 | 1272 | |||
BC032329.1 | - | 8 | 1345 | 5165263 | ||
KU178011.1 | - | 8 | 1301 | |||
M14565.1 | - | 8 | 1345 | |||
M28253.1 | - | 4 | 455 |