Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005417.2 | NP_001005417.1 | 6 | 1115 | ||
NM_003780.4 | NP_003771.1 | 6 | 1315 | |||
NM_030587.2 | NP_085076.2 | 6 | 1116 | |||
XM_017002716.1 | XP_016858205.1 | 6 | 1073 | |||
XM_017002717.1 | XP_016858206.1 | 6 | 1276 | |||
GenBank mRNA | AB024434.1 | - | 5 | 961 | ||
AK074533.1 | - | 4 | 613 | |||
AK095873.1 | - | 6 | 1743 | |||
AK293418.1 | - | 6 | 1065 | |||
AL137647.1 | - | 4 | 510 | |||
BC096821.1 | - | 6 | 1083 | 30558762 | ||
Y12510.1 | - | 5 | 953 |