Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005502.3 | NP_005493.2 | 7, 8 | 1123 | ||
XM_005251773.2 | XP_005251830.1 | 7, 8 | 1032 | |||
XM_005251776.2 | XP_005251833.1 | 6, 7 | 872 | |||
XM_011518339.2 | XP_011516641.1 | 8, 9 | 1107 | |||
XM_011518340.2 | XP_011516642.1 | 9, 10 | 1322 | |||
XM_011518341.2 | XP_011516643.1 | 8, 9 | 1107 | |||
XM_011518342.2 | XP_011516644.1 | 5, 6 | 810 | |||
XM_011518344.2 | XP_011516646.1 | 8, 9 | 1107 | |||
XM_017014378.1 | XP_016869867.1 | 8, 9 | 1305 | |||
XM_017014379.1 | XP_016869868.1 | 8, 9 | 1713 | |||
XM_017014380.1 | XP_016869869.1 | 8, 9 | 1039 | |||
XM_017014381.1 | XP_016869870.1 | 8, 9 | 1105 | |||
XM_017014382.1 | XP_016869871.1 | 6, 7 | 1570 | |||
GenBank mRNA | AB055982.1 | - | 6, 7 | 719 | ||
AB445477.1 | - | 6, 7 | 719 | |||
AF165281.1 | - | 5, 6 | 659 | |||
AF285167.1 | - | 7, 8 | 1009 | |||
AJ012376.1 | - | 5, 6 | 659 | |||
BC146856.1 | - | 7, 8 | 933 | 9056784 |