Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012413.3 | NP_036545.1 | 5, 6 | 947 | ||
GenBank mRNA | AK290605.1 | - | 5, 6 | 929 | ||
BC020456.1 | - | 5, 6 | 848 | |||
BC036721.1 | - | 5, 6 | 826 | 5224222 | ||
BC047756.2 | - | 5, 6 | 924 | 6058335 | ||
X67731.1 | - | 4, 5 | 629 | |||
X71125.1 | - | 5, 6 | 836 |