Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307986.1 | NP_001294915.1 | 4, 5 | 1074 | ||
NM_001307987.1 | NP_001294916.1 | 2, 3 | 873 | |||
NM_013305.5 | NP_037437.2 | 3, 4 | 966 | |||
XM_017025715.1 | XP_016881204.1 | 5, 6 | 462 | |||
GenBank mRNA | AK056270.1 | - | 3, 4 | 966 | ||
AK293608.1 | - | 4, 5 | 714 | |||
AK298881.1 | - | 2, 3 | 514 | |||
BC108910.1 | - | 3, 4 | 516 | 40011279 | ||
BC108911.1 | - | 3, 4 | 516 | 40011281 | ||
CR457037.1 | - | 3, 4 | 305 | |||
U91641.1 | - | 3, 4 | 352 |