Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307986.1 | NP_001294915.1 | 8 | 2230 | ||
NM_001307987.1 | NP_001294916.1 | 6 | 2029 | |||
NM_013305.5 | NP_037437.2 | 7 | 2122 | |||
XM_017025715.1 | XP_016881204.1 | 9 | 1618 | |||
XM_017025716.1 | XP_016881205.1 | 7 | 1796 | |||
GenBank mRNA | AK056270.1 | - | 7 | 2122 | ||
AK094273.1 | - | 6 | 1635 | |||
AK130126.1 | - | 9 | 1625 | |||
AK293608.1 | - | 8 | 1870 | |||
BC108910.1 | - | 7 | 1672 | 40011279 | ||
BC108911.1 | - | 7 | 1672 | 40011281 | ||
U91641.1 | - | 7 | 1517 |