Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013338.4 | NP_037470.1 | 4 | 364 | ||
XM_017020552.1 | XP_016876041.1 | 3 | 302 | |||
GenBank mRNA | AF102850.1 | - | 4 | 372 | ||
AF161498.1 | - | 4 | 313 | |||
AF183413.1 | - | 4 | 323 | |||
AK299126.1 | - | 4 | 333 | |||
BC012531.1 | - | 4 | 316 | 4273297 | ||
BF668393.1 | - | 338 | ||||
BG482604.1 | - | 4 | 349 | |||
BP246765.1 | - | 4 | 364 | |||
CB988638.1 | - | 361 |