Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014475.3 | NP_055290.1 | 7 | 951 | ||
XM_005258748.4 | XP_005258805.1 | 6 | 1348 | |||
XM_017026598.1 | XP_016882087.1 | 7 | 1511 | |||
GenBank mRNA | AB021933.1 | - | 7 | 911 | ||
BC032730.1 | - | 7 | 962 | 5530225 |