Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317395.1 | NP_001304324.1 | 3 | 536 | ||
NM_001317399.1 | NP_001304328.1 | 3 | 536 | |||
NM_031954.4 | NP_114160.1 | 3 | 536 | |||
NR_133898.1 | 2 | 322 | ||||
GenBank mRNA | AF113208.1 | - | 3 | 281 | ||
AK027543.1 | - | 3 | 342 | |||
AK056912.1 | - | 3 | 1941 | |||
AK222548.1 | - | 3 | 292 | |||
AK309783.1 | - | 2 | 151 | |||
AY205299.1 | - | 3 | 342 | |||
AY597809.1 | - | 2 | 254 | |||
BC040062.1 | - | 3 | 289 | 5494016 | ||
BX333975.2 | - | 3 | 286 | |||
CX785497.1 | - | 2 | 65 | |||
DA440672.1 | - | 3 | 536 |