Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286668.1 | NP_001273597.1 | 3 | 339 | ||
NM_001286669.1 | NP_001273598.1 | 3 | 490 | |||
NM_001286671.1 | NP_001273600.1 | 4 | 611 | |||
NM_001286672.1 | NP_001273601.1 | 4 | 611 | |||
NM_001323890.1 | NP_001310819.1 | 4 | 611 | |||
NM_001323892.1 | NP_001310821.1 | 4 | 611 | |||
NM_001323893.1 | NP_001310822.1 | 3 | 490 | |||
NM_001323899.1 | NP_001310828.1 | 4 | 611 | |||
NM_181806.3 | NP_861522.2 | 4 | 611 | |||
XM_017007740.1 | XP_016863229.1 | 4 | 572 | |||
XM_017007741.1 | XP_016863230.1 | 5 | 687 | |||
XM_017007742.1 | XP_016863231.1 | 2 | 387 | |||
XM_017007743.1 | XP_016863232.1 | 4 | 572 | |||
XM_017007744.1 | XP_016863233.1 | 2 | 387 | |||
GenBank mRNA | AF161436.1 | - | 4 | 560 | ||
AF516672.1 | - | 3 | 254 | |||
AK303733.1 | - | 3 | 443 | |||
AK304309.1 | - | 4 | 602 | |||
AK316296.1 | - | 4 | 592 | |||
AY314787.1 | - | 4 | 574 | |||
AY422212.1 | - | 4 | 526 | |||
BC141824.1 | - | 4 | 569 | 40148174 | ||
BC142709.1 | - | 4 | 547 | 40148820 | ||
BX640635.1 | - | 2 | 138 | |||
BX648853.1 | - | 4 | 603 |