Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003104.5 | NP_003095.2 | 3, 4 | 450 | ||
GenBank mRNA | AK312444.1 | - | 3, 4 | 423 | ||
BC021085.2 | - | 3, 4 | 400 | 2989137 | ||
BC025295.1 | - | 3, 4 | 348 | 5428022 | ||
DB031906.1 | - | 3, 4 | 450 | |||
FJ224316.1 | - | 3, 4 | 400 | |||
L29008.1 | - | 3, 4 | 407 | |||
U07361.1 | - | 3, 4 | 411 |