Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007272.2 | NP_009203.2 | 7, 8 | 820 | ||
XM_011540550.1 | XP_011538852.1 | 6, 7 | 674 | |||
GenBank mRNA | AK289447.1 | - | 7, 8 | 819 | ||
BC015118.1 | - | 7, 8 | 826 | 3950170 | ||
BC059779.1 | - | 6, 7 | 653 | |||
CR749296.1 | - | 4, 5 | 484 | |||
S82198.1 | - | 7, 8 | 797 | |||
Y13697.1 | - | 6, 7 | 749 |