Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014331.3 | NP_055146.1 | 1 | 469 | ||
XM_011531802.2 | XP_011530104.1 | 1 | 538 | |||
GenBank mRNA | AB026891.1 | - | 1 | 424 | ||
AB040875.1 | - | 1 | 324 | |||
AF200708.1 | - | 1 | 434 | |||
AF252872.1 | - | 1 | 420 | |||
AJ277882.1 | - | 1 | 189 | |||
AK290359.1 | - | 1 | 469 | |||
AK314855.1 | - | 1 | 363 | |||
BC012087.1 | - | 1 | 349 | 4562994 | ||
CD639041.1 | - | 1 | 472 |