Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PISD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326411.1 | NP_001313340.1 | 5 | 942 | ||
NM_001326412.1 | NP_001313341.1 | 5 | 879 | |||
NM_001326413.1 | NP_001313342.1 | 5 | 1028 | |||
NM_001326414.1 | NP_001313343.1 | 6 | 1108 | |||
NM_001326415.1 | NP_001313344.1 | 6 | 1282 | |||
NM_001326416.1 | NP_001313345.1 | 8 | 1471 | |||
NM_001326417.1 | NP_001313346.1 | 7 | 1362 | |||
NM_001326418.1 | NP_001313347.1 | 4 | 862 | |||
NM_001326419.1 | NP_001313348.1 | 7 | 1362 | |||
NM_001326420.1 | NP_001313349.1 | 4 | 862 | |||
NM_001326421.1 | NP_001313350.1 | 5 | 942 | |||
NM_014338.3 | NP_055153.1 | 6 | 914 | |||
NM_178022.1 | NP_821141.1 | 5 | 823 | |||
GenBank mRNA | AK298474.1 | - | 5 | 823 | ||
AL050371.1 | - | 4 | 769 | |||
BC001482.2 | - | 6 | 914 | 3508281 | ||
CR456540.1 | - | 5 | 592 |