Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143679.1 | NP_001137151.1 | 4 | 453 | ||
NM_001143680.1 | NP_001137152.1 | 4 | 453 | |||
NM_001143681.1 | NP_001137153.1 | 3 | 324 | |||
NM_015917.2 | NP_057001.1 | 4 | 453 | |||
GenBank mRNA | AF068287.1 | - | 4 | 440 | ||
AF070657.1 | - | 4 | 411 | |||
AF087849.1 | - | 4 | 425 | |||
AK000640.1 | - | 4 | 411 | |||
AK095263.1 | - | 3 | 710 | |||
AK289570.1 | - | 4 | 386 | |||
AK289580.1 | - | 4 | 391 | |||
AK295592.1 | - | 3 | 312 | |||
AK299968.1 | - | 4 | 371 | |||
AL136938.1 | - | 4 | 439 | |||
AY520571.1 | - | 4 | 422 | |||
BC001231.1 | - | 4 | 400 | 3460150 | ||
BC050715.1 | - | 4 | 373 | 6061265 | ||
BC063425.1 | - | 4 | 382 | 5218251 | ||
BC110600.1 | - | 2 | 209 | |||
BQ436828.1 | - | 4 | 453 | |||
CR533445.1 | - | 4 | 368 |