Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204284.1 | NP_001191213.1 | 6 | 871 | ||
NM_006247.3 | NP_006238.1 | 7 | 937 | |||
XM_017026935.1 | XP_016882424.1 | 7 | 932 | |||
GenBank mRNA | AB209615.1 | - | 2 | 2414 | ||
AK094241.1 | - | 2 | 1143 | |||
AK223222.1 | - | 7 | 844 | |||
AK293402.1 | - | 8 | 795 | |||
AK293475.1 | - | 3 | 452 | |||
AK297095.1 | - | 8 | 783 | |||
AK312683.1 | - | 7 | 844 | |||
BC000750.2 | - | 7 | 830 | |||
BC001831.2 | - | 7 | 830 | |||
BC001970.1 | - | 7 | 894 | 3459309 | ||
BT007275.1 | - | 7 | 834 | |||
U25174.1 | - | 7 | 812 | |||
X89416.1 | - | 7 | 817 |