Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007352.3 | NP_031378.1 | 7 | 847 | ||
GenBank mRNA | AK092382.1 | - | 9 | 642 | ||
AK315798.1 | - | 7 | 716 | |||
BC005216.1 | - | 7 | 721 | 3949903 | ||
M16630.1 | - | 7 | 692 | |||
M18692.1 | - | 7 | 719 |