Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002536.2 | NP_002527.1 | 6 | 2360 | ||
GenBank mRNA | AK294776.1 | - | 6 | 2258 | ||
AK297924.1 | - | 3 | 1765 | |||
AK299561.1 | - | 1 | 1288 | |||
AK316199.1 | - | 4 | 2040 | |||
BC026050.1 | - | 1 | 1283 | |||
BC036942.1 | - | 3 | 1789 | |||
BC052609.1 | - | 5 | 2150 | |||
BC082255.1 | - | 4 | 2260 | |||
BG328213.1 | - | 1 | 470 | |||
BI088539.1 | - | 1 | 395 | |||
L08240.1 | - | 5 | 2139 |