Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145290.3 | NP_660333.2 | 10, 11 | 1719 | ||
XM_005248137.4 | XP_005248194.1 | 10, 11 | 1723 | |||
XM_011513811.2 | XP_011512113.1 | 5, 6 | 903 | |||
XM_017007834.1 | XP_016863323.1 | 10, 11 | 1245 | |||
GenBank mRNA | AK095866.1 | - | 5, 6 | 901 | ||
AK299168.1 | - | 7, 8 | 1006 | |||
AL832257.1 | - | 2, 3 | 188 | |||
AY181243.1 | - | 8, 9 | 1074 | |||
AY358306.1 | - | 10, 11 | 1662 | |||
BC026009.1 | - | 5, 6 | 803 | |||
BC035645.1 | - | 10, 11 | 1730 |
Set Membership: |
G-protein coupled receptor |