Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145290.3 | NP_660333.2 | 14, 15 | 2502 | ||
XM_005248137.4 | XP_005248194.1 | 14, 15 | 2506 | |||
XM_011513811.2 | XP_011512113.1 | 9, 10 | 1686 | |||
XM_017007834.1 | XP_016863323.1 | 14, 15 | 2028 | |||
GenBank mRNA | AK027494.1 | - | 3, 4 | 555 | ||
AK095866.1 | - | 9, 10 | 1684 | |||
AK299168.1 | - | 11, 12 | 1789 | |||
AL832257.1 | - | 6, 7 | 971 | |||
AY181243.1 | - | 12, 13 | 1803 | |||
BC035645.1 | - | 14, 15 | 2513 |
Set Membership: |
G-protein coupled receptor |