Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014366.4 | NP_055181.3 | 3 | 352 | ||
NM_206825.1 | NP_996561.1 | 3 | 449 | |||
NM_206826.1 | NP_996562.1 | 3 | 302 | |||
GenBank mRNA | AF191018.1 | - | 3 | 349 | ||
AI028402.1 | - | 3 | 352 | |||
AK027514.1 | - | 3 | 353 | |||
AK027516.1 | - | 3 | 202 | |||
AK315484.1 | - | 3 | 265 | |||
AY825265.1 | - | 2 | 179 | |||
BC001024.2 | - | 3 | 200 | 3342771 | ||
CF272475.1 | - | 3 | 221 |