Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143679.1 | NP_001137151.1 | 1, 2 | 165 | ||
NM_001143680.1 | NP_001137152.1 | 1, 2 | 165 | |||
NM_001143681.1 | NP_001137153.1 | 1, 2 | 165 | |||
NM_015917.2 | NP_057001.1 | 1, 2 | 165 | |||
GenBank mRNA | AF068287.1 | - | 1, 2 | 152 | ||
AF070657.1 | - | 1, 2 | 123 | |||
AF087849.1 | - | 1, 2 | 137 | |||
AK000640.1 | - | 1, 2 | 123 | |||
AK095263.1 | - | 1, 2 | 107 | |||
AK289570.1 | - | 1, 2 | 98 | |||
AK289580.1 | - | 1, 2 | 103 | |||
AK295592.1 | - | 1, 2 | 153 | |||
AK299968.1 | - | 1, 2 | 83 | |||
AL136938.1 | - | 1, 2 | 151 | |||
AY520571.1 | - | 1, 2 | 134 | |||
BC001231.1 | - | 1, 2 | 112 | 3460150 | ||
BC050715.1 | - | 1, 2 | 85 | 6061265 | ||
BC063425.1 | - | 1, 2 | 94 | 5218251 | ||
BQ436828.1 | - | 1, 2 | 165 | |||
CR533445.1 | - | 1, 2 | 80 |