Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FETUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014375.2 | NP_055190.2 | 2 | 406 | ||
XM_005247351.3 | XP_005247408.1 | 2 | 439 | |||
XM_011512681.2 | XP_011510983.1 | 3 | 707 | |||
XM_011512682.2 | XP_011510984.1 | 2 | 443 | |||
GenBank mRNA | AB017551.1 | - | 2 | 404 | ||
AF534529.1 | - | 2 | 406 | |||
AJ242928.1 | - | 2 | 406 | |||
AK309647.1 | - | 2 | 171 | |||
AK315309.1 | - | 3 | 568 | |||
BC069670.1 | - | 2 | 403 | 7262597 | ||
BC069820.1 | - | 2 | 403 | 7262609 | ||
BC074734.2 | - | 2 | 345 | 30915188 | ||
BC114616.1 | - | 2 | 396 | 40032724 |