Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBE3A SIRNA Assays ›
Gene Symbol:
UBE3A
Gene Name:
ubiquitin protein ligase E3A
Gene Aliases:
ANCR, AS, E6-AP, EPVE6AP, HPVE6A
Chromosome Location:
Chr.15: 25337234 - 25439086 on Build GRCh38
UniGene ID:
Hs.598862
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
213353
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBE3A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000462.3
NP_000453.2
7
1321
NM_130838.1
NP_570853.1
3
536
NM_130839.2
NP_570854.1
6
1256
XM_005268267.4
XP_005268324.1
8
1623
XM_005268268.4
XP_005268325.1
7
1510
XM_005268269.4
XP_005268326.1
8
1354
XM_005268270.4
XP_005268327.1
7
1311
XM_005268271.4
XP_005268328.1
4
1130
XM_006720675.3
XP_006720738.1
8
1575
XM_006720676.3
XP_006720739.1
5
820
XM_011521994.2
XP_011520296.1
8
1512
XM_011521995.2
XP_011520297.1
8
1186
XM_017022544.1
XP_016878033.1
9
1548
XM_017022545.1
XP_016878034.1
8
1507
XM_017022546.1
XP_016878035.1
8
1415
XM_017022547.1
XP_016878036.1
7
1447
XM_017022548.1
XP_016878037.1
7
1442
XM_017022549.1
XP_016878038.1
7
1249
XM_017022550.1
XP_016878039.1
7
1401
XM_017022551.1
XP_016878040.1
8
1452
XM_017022552.1
XP_016878041.1
8
1142
XM_017022553.1
XP_016878042.1
8
1457
XM_017022554.1
XP_016878043.1
4
1461
XM_017022555.1
XP_016878044.1
6
1230
XM_017022556.1
XP_016878045.1
7
1379
XM_017022557.1
XP_016878046.1
6
1314
XM_017022558.1
XP_016878047.1
7
1510
XM_017022559.1
XP_016878048.1
4
1130
GenBank mRNA
AB194232.1
-
5
632
AF273050.1
-
4
591
AK056239.1
-
6
1224
AK130491.1
-
2
265
AK292514.1
-
6
866
BC002582.2
-
6
1209
3160225
L07557.1
-
3
531
U84404.1
-
8
1078
X98031.1
-
4
561
X98032.1
-
2
492
X98033.1
-
4
552
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
proteolysis
ubiquitin-dependent protein catabolic process
brain development
positive regulation of phosphatidylinositol 3-kinase signaling
viral process
androgen receptor signaling pathway
positive regulation of protein ubiquitination
sperm entry
regulation of circadian rhythm
positive regulation of transcription from RNA polymerase II promoter
protein autoubiquitination
prostate gland growth
protein K48-linked ubiquitination
regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
transcription coactivator activity
ubiquitin-protein transferase activity
protein binding
ligase activity
metal ion binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-zrdb5:80/100.66.79.80:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline